6-Phenyl-2-pyrrolidin-2-ylmethyl-2H-pyridazin-3-one hydrochloride

Supplier: Matrix Scientific

067815-250MG
101934-748EA 476.92 USD
101934-748
6-Phenyl-2-pyrrolidin-2-ylmethyl-2H-pyridazin-3-one hydrochloride
6-Phenyl-2-pyrrolidin-2-ylmethyl-2H-pyridazin-3-one hydrochloride

Hazard: Irritant

Formula: C₁₅H₁₈ClN₃O
MDL Number: MFCD16621937
CAS Number: 1229625-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR