5,6,7,8-Tetrahydro-2H-cinnolin-3-one

Supplier: Matrix Scientific

Synonyms: 5,6,7,8-Tetrahydrocinnolin-3-ol

067812-500MG
101934-742EA 623.08 USD
101934-742
5,6,7,8-Tetrahydro-2H-cinnolin-3-one
5,6,7,8-Tetrahydro-2H-cinnolin-3-one

Hazard: Irritant

Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
MDL Number: MFCD00828986
CAS Number: 5468-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR