2-(2-Amino-1-methylethyl)-6-thiophen-2-yl-2H-pyridazin-3-one hydrochloride

Supplier: Matrix Scientific

067807-250MG
101934-732EA 476.92 USD
101934-732
2-(2-Amino-1-methylethyl)-6-thiophen-2-yl-2H-pyridazin-3-one hydrochloride
2-(2-Amino-1-methylethyl)-6-thiophen-2-yl-2H-pyridazin-3-one hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄ClN₃OS
MDL Number: MFCD21605991
CAS Number: 1361112-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR