3-(2-Aminoethyl)-3H-benzooxazol-2-one hydrochloride

Supplier: Matrix Scientific

067806-250MG
101934-730EA 476.92 USD
101934-730
3-(2-Aminoethyl)-3H-benzooxazol-2-one hydrochloride
3-(2-Aminoethyl)-3H-benzooxazol-2-one hydrochloride

Hazard: Irritant

Formula: C₉H₁₁ClN₂O₂
MDL Number: MFCD17977099
CAS Number: 1258640-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR