2-(R)-1-Piperidin-3-ylmethyl-6-thiophen-2-yl-2H-pyridazin-3-one hydrochloride

Supplier: Matrix Scientific

067798-250MG
101934-714EA 558.46 USD
101934-714
2-(R)-1-Piperidin-3-ylmethyl-6-thiophen-2-yl-2H-pyridazin-3-one hydrochloride
2-(R)-1-Piperidin-3-ylmethyl-6-thiophen-2-yl-2H-pyridazin-3-one hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₈ClN₃OS
MDL Number: MFCD16621989
CAS Number: 1229622-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR