2-Methylpyrrolidine-1,2-dicarboxylic acid-1-tert-butyl ester
Supplier: Matrix Scientific
Synonyms:
1-Boc-2-methyl-2-pyrrolidinecarboxylic acid
, 1-Boc-2-methyl-DL-proline
067785-250MG
101934-688EA
476.92
USD
101934-688
2-Methylpyrrolidine-1,2-dicarboxylic acid-1-tert-butyl ester
2-Methylpyrrolidine-1,2-dicarboxylic acid-1-tert-butyl ester
Hazard: Irritant
Formula:
C₁₁H₁₉NO₄ MW: 229.28 g/mol |
MDL Number:
MFCD03095494 CAS Number: 203869-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...