1-Pyrimidin-2-ylpiperidine-3-carboxylic acid
Supplier: Matrix Scientific
067752-500MG
101934-622EA
623.08
USD
101934-622
1-Pyrimidin-2-ylpiperidine-3-carboxylic acid
1-Pyrimidin-2-ylpiperidine-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₁₃N₃O₂ |
MDL Number:
MFCD06255172 CAS Number: 890013-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...