3-Methyl-5-pyrrolidin-2-yl[1,2,4]oxadiazole hydrochloride

Supplier: Matrix Scientific

067739-500MG
101934-596EA 623.08 USD
101934-596
3-Methyl-5-pyrrolidin-2-yl[1,2,4]oxadiazole hydrochloride
3-Methyl-5-pyrrolidin-2-yl[1,2,4]oxadiazole hydrochloride

Hazard: Irritant

Formula: C₇H₁₂ClN₃O
MDL Number: MFCD21605983
CAS Number: 1361113-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR