1-Cyclohexylcarbamoylmethyl-6-oxo-1,6-dihydropyridine-3-carboxylic acid

Supplier: Matrix Scientific

067737-500MG
101934-592EA 623.08 USD
101934-592
1-Cyclohexylcarbamoylmethyl-6-oxo-1,6-dihydropyridine-3-carboxylic acid
1-Cyclohexylcarbamoylmethyl-6-oxo-1,6-dihydropyridine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₄
MDL Number: MFCD13366974
CAS Number: 1041535-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR