(6-Methyl-1',2',3',4',5',6'-hexahydro[2,4']bipyridinyl-4-yl)pyrimidin-2-ylamine

Supplier: Matrix Scientific

067733-250MG
101934-584EA 476.92 USD
101934-584
(6-Methyl-1',2',3',4',5',6'-hexahydro[2,4']bipyridinyl-4-yl)pyrimidin-2-ylamine
(6-Methyl-1',2',3',4',5',6'-hexahydro[2,4']bipyridinyl-4-yl)pyrimidin-2-ylamine

Hazard: Irritant

Formula: C₁₅H₁₉N₅
MDL Number: MFCD19691431
CAS Number: 1316222-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR