(2-Methyl-6-pyrrolidin-2-ylpyridin-4-yl)pyrazin-2-ylamine

Supplier: Matrix Scientific

067732-250MG
101934-582EA 558.46 USD
101934-582
(2-Methyl-6-pyrrolidin-2-ylpyridin-4-yl)pyrazin-2-ylamine
(2-Methyl-6-pyrrolidin-2-ylpyridin-4-yl)pyrazin-2-ylamine

Hazard: Irritant

Formula: C₁₄H₁₇N₅
MDL Number: MFCD21605979
CAS Number: 1361118-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR