6-Oxo-1-(thiazol-2-ylcarbamoylmethyl)-1,6-dihydropyridine-3-carboxylic acid

Supplier: Matrix Scientific

067730-500MG
101934-578EA 623.08 USD
101934-578
6-Oxo-1-(thiazol-2-ylcarbamoylmethyl)-1,6-dihydropyridine-3-carboxylic acid
6-Oxo-1-(thiazol-2-ylcarbamoylmethyl)-1,6-dihydropyridine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₉N₃O₄S
MW: 279.28 g/mol
MDL Number: MFCD16621918
CAS Number: 1361112-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR