1-Dimethylcarbamoylmethyl-6-oxo-1,6-dihydropyridine-3-carboxylic acid

Supplier: Matrix Scientific

067727-500MG
101934-572EA 623.08 USD
101934-572
1-Dimethylcarbamoylmethyl-6-oxo-1,6-dihydropyridine-3-carboxylic acid
1-Dimethylcarbamoylmethyl-6-oxo-1,6-dihydropyridine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₄
MDL Number: MFCD11115833
CAS Number: 1036611-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR