3-Carbamoylpiperazine-1-carboxylic acid tert-butyl ester

Supplier: Matrix Scientific

Synonyms: 1-Boc-3-Carbamoylpiperazine

067726-250MG
101934-570EA 769.23 USD
101934-570
3-Carbamoylpiperazine-1-carboxylic acid tert-butyl ester
3-Carbamoylpiperazine-1-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₀H₁₉N₃O₃
MW: 229.28 g/mol
MDL Number: MFCD09608050
CAS Number: 112257-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR