1'-Methanesulfonyl[2,4']bipiperidinyl hydrochloride

Supplier: Matrix Scientific

067724-250MG
101934-566EA 630.77 USD
101934-566
1'-Methanesulfonyl[2,4']bipiperidinyl hydrochloride
1'-Methanesulfonyl[2,4']bipiperidinyl hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₃ClN₂O₂S
MDL Number: MFCD21605975
CAS Number: 1361113-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR