1-Methanesulfonyl-3-methylpiperidine-3-carboxylic acid

Supplier: Matrix Scientific

067716-250MG
101934-550EA 476.92 USD
101934-550
1-Methanesulfonyl-3-methylpiperidine-3-carboxylic acid
1-Methanesulfonyl-3-methylpiperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₈H₁₅NO₄S
MDL Number: MFCD18273395
CAS Number: 1316226-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR