1-(2-Methoxyacetyl)-4-methylpiperidine-4-carboxylic acid

Supplier: Matrix Scientific

067713-250MG
101934-544EA 476.92 USD
101934-544
1-(2-Methoxyacetyl)-4-methylpiperidine-4-carboxylic acid
1-(2-Methoxyacetyl)-4-methylpiperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₇NO₄
MDL Number: MFCD18271887
CAS Number: 1316220-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR