1-(2-Methoxyacetyl)-3-methylpiperidine-3-carboxylic acid

Supplier: Matrix Scientific

067705-500MG
101934-528EA 623.08 USD
101934-528
1-(2-Methoxyacetyl)-3-methylpiperidine-3-carboxylic acid
1-(2-Methoxyacetyl)-3-methylpiperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₇NO₄
MDL Number: MFCD18273183
CAS Number: 1316221-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR