[2,4']Bipiperidinyl-1-yl(4-methylthiazol-5-yl)methanone hydrochloride

Supplier: Matrix Scientific

067699-250MG
101934-516EA 476.92 USD
101934-516
[2,4']Bipiperidinyl-1-yl(4-methylthiazol-5-yl)methanone hydrochloride
[2,4']Bipiperidinyl-1-yl(4-methylthiazol-5-yl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClN₃OS
MDL Number: MFCD21605961
CAS Number: 1361112-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR