Morpholin-4-yl(6-pyrrolidin-2-ylpyridin-3-yl)methanonehydrochloride
Supplier: Matrix Scientific
067697-250MG
101934-512EA
558.46
USD
101934-512
Morpholin-4-yl(6-pyrrolidin-2-ylpyridin-3-yl)methanonehydrochloride
Morpholin-4-yl(6-pyrrolidin-2-ylpyridin-3-yl)methanonehydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₀ClN₃O₂ MW: 297.79 g/mol |
MDL Number:
MFCD21605959 CAS Number: 1361114-83-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...