6-Pyrrolidin-2-yl-N-thiazol-2-ylnicotinamide hydrochloride

Supplier: Matrix Scientific

067696-500MG
101934-510EA 623.08 USD
101934-510
6-Pyrrolidin-2-yl-N-thiazol-2-ylnicotinamide hydrochloride
6-Pyrrolidin-2-yl-N-thiazol-2-ylnicotinamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₅ClN₄OS
MW: 310.81 g/mol
MDL Number: MFCD21605958
CAS Number: 1361114-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR