1'-(2-Fluorobenzyl)-1',2',3',4',5',6'-hexahydro[2,3']bipyridinyl-5-carboxylic acid hydrochloride

Supplier: Matrix Scientific

067693-250MG
101934-504EA 476.92 USD
101934-504
1'-(2-Fluorobenzyl)-1',2',3',4',5',6'-hexahydro[2,3']bipyridinyl-5-carboxylic acid hydrochloride
1'-(2-Fluorobenzyl)-1',2',3',4',5',6'-hexahydro[2,3']bipyridinyl-5-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₁₈H₂₀ClFN₂O₂
MDL Number: MFCD21605955
CAS Number: 1361116-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR