2-(4-Aminopyrazol-1-yl)-N,N-dimethylacetamide hydrochloride

Supplier: Matrix Scientific

067690-250MG
101934-498EA 476.92 USD
101934-498
2-(4-Aminopyrazol-1-yl)-N,N-dimethylacetamide hydrochloride
2-(4-Aminopyrazol-1-yl)-N,N-dimethylacetamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₃ClN₄O
MDL Number: MFCD21605952
CAS Number: 1361112-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR