2-Methyl-6-pyrrolidin-2-ylpyrimidin-4-ol dihydrochloride

Supplier: Matrix Scientific

067689-500MG
101934-496EA 623.08 USD
101934-496
2-Methyl-6-pyrrolidin-2-ylpyrimidin-4-ol dihydrochloride
2-Methyl-6-pyrrolidin-2-ylpyrimidin-4-ol dihydrochloride

Hazard: Irritant

Formula: C₉H₁₅Cl₂N₃O
MDL Number: MFCD16621923
CAS Number: 1229623-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR