2-(4-Aminopyrazol-1-yl)-N-[2-(2-fluorophenyl)ethyl]acetamide hydrochloride

Supplier: Matrix Scientific

067686-250MG
101934-490EA 476.92 USD
101934-490
2-(4-Aminopyrazol-1-yl)-N-[2-(2-fluorophenyl)ethyl]acetamide hydrochloride
2-(4-Aminopyrazol-1-yl)-N-[2-(2-fluorophenyl)ethyl]acetamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₆ClFN₄O
MDL Number: MFCD21605949
CAS Number: 1361115-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR