1-(2-Methyl-7-morpholin-2-ylpyrazolo[1,5-a]pyrimidin-3-ylmethyl)piperidin-4-oldihydrochloride

Supplier: Matrix Scientific

067674-250MG
101934-466EA 687.69 USD
101934-466
1-(2-Methyl-7-morpholin-2-ylpyrazolo[1,5-a]pyrimidin-3-ylmethyl)piperidin-4-oldihydrochloride
1-(2-Methyl-7-morpholin-2-ylpyrazolo[1,5-a]pyrimidin-3-ylmethyl)piperidin-4-oldihydrochloride

Hazard: Irritant

Formula: C₁₇H₂₇Cl₂N₅O₂
MDL Number: MFCD21605940
CAS Number: 1361113-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR