(6-Methyl-1',2',3',4',5',6'-hexahydro[2,4']bipyridinyl-4-yl)pyridin-2-ylamine
Supplier: Matrix Scientific
067666-250MG
101934-450EA
476.92
USD
101934-450
(6-Methyl-1',2',3',4',5',6'-hexahydro[2,4']bipyridinyl-4-yl)pyridin-2-ylamine
(6-Methyl-1',2',3',4',5',6'-hexahydro[2,4']bipyridinyl-4-yl)pyridin-2-ylamine
Hazard: Irritant
Formula:
C₁₆H₂₀N₄ |
MDL Number:
MFCD19691439 CAS Number: 1316222-73-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...