3-Azepan-2-ylmethylbenzo[b]thiophene-2-carboxylic acid(2-hydroxyethyl)amide hydrochloride

Supplier: Matrix Scientific

067659-250MG
101934-436EA 476.92 USD
101934-436
3-Azepan-2-ylmethylbenzo[b]thiophene-2-carboxylic acid(2-hydroxyethyl)amide hydrochloride
3-Azepan-2-ylmethylbenzo[b]thiophene-2-carboxylic acid(2-hydroxyethyl)amide hydrochloride

Hazard: Irritant

Formula: C₁₈H₂₅ClN₂O₂S
MDL Number: MFCD21605936
CAS Number: 1361112-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR