2-(5-Bromopyridin-2-yl)-4-methanesulfonylmorpholine

Supplier: Matrix Scientific

067658-250MG
101934-434EA 638.46 USD
101934-434
2-(5-Bromopyridin-2-yl)-4-methanesulfonylmorpholine
2-(5-Bromopyridin-2-yl)-4-methanesulfonylmorpholine

Hazard: Irritant

Formula: C₁₀H₁₃BrN₂O₃S
MW: 321.2 g/mol
MDL Number: MFCD19691607
CAS Number: 1316223-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR