Pyridin-2-yl-(6-pyrrolidin-2-ylpyridin-3-yl)amine dihydrochloride

Supplier: Matrix Scientific

067650-250MG
101934-418EA 638.46 USD
101934-418
Pyridin-2-yl-(6-pyrrolidin-2-ylpyridin-3-yl)amine dihydrochloride
Pyridin-2-yl-(6-pyrrolidin-2-ylpyridin-3-yl)amine dihydrochloride

Hazard: Irritant

Formula: C₁₄H₁₈Cl₂N₄
MW: 313.23 g/mol
MDL Number: MFCD21605931
CAS Number: 1361116-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR