N-(1-Ethyl-1H-pyrazol-3-yl)-2-piperidin-4-ylacetamide hydrochloride
Supplier: Matrix Scientific
067646-500MG
101934-410EA
623.08
USD
101934-410
N-(1-Ethyl-1H-pyrazol-3-yl)-2-piperidin-4-ylacetamide hydrochloride
N-(1-Ethyl-1H-pyrazol-3-yl)-2-piperidin-4-ylacetamide hydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₁ClN₄O MW: 272.78 g/mol |
MDL Number:
MFCD21605928 CAS Number: 1361115-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...