4-Azetidin-3-ylpyrimidin-2-ylaminetris(trifluoroacetic acid salt)

Supplier: Matrix Scientific

067643-250MG
101934-404EA 692.31 USD
101934-404
4-Azetidin-3-ylpyrimidin-2-ylaminetris(trifluoroacetic acid salt)
4-Azetidin-3-ylpyrimidin-2-ylaminetris(trifluoroacetic acid salt)

Hazard: Irritant

Formula: C₁₃H₁₃F₉N₄O₆
MDL Number: MFCD21605925
CAS Number: 1361113-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR