Pyrrolidine-3-carboxylic acid (2-methylbenzothiazol-6-yl)amide hydrochloride
Supplier: Matrix Scientific
067639-500MG
101934-396EA
623.08
USD
101934-396
Pyrrolidine-3-carboxylic acid (2-methylbenzothiazol-6-yl)amide hydrochloride
Pyrrolidine-3-carboxylic acid (2-methylbenzothiazol-6-yl)amide hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₆ClN₃OS MW: 297.81 g/mol |
MDL Number:
MFCD21605922 CAS Number: 1361116-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...