2-Methyl-4-piperidin-2-ylpyrimidine dihydrochloride

Supplier: Matrix Scientific

067638-250MG
101934-394EA 476.92 USD
101934-394
2-Methyl-4-piperidin-2-ylpyrimidine dihydrochloride
2-Methyl-4-piperidin-2-ylpyrimidine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₇Cl₂N₃
MDL Number: MFCD21605921
CAS Number: 1361112-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR