4-Pyrrolidin-2-yl-6-trifluoromethylpyrimidin-2-ylamine hydrochloride

Supplier: Matrix Scientific

067636-250MG
101934-390EA 558.46 USD
101934-390
4-Pyrrolidin-2-yl-6-trifluoromethylpyrimidin-2-ylamine hydrochloride
4-Pyrrolidin-2-yl-6-trifluoromethylpyrimidin-2-ylamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₂ClF₃N₄
MW: 268.67 g/mol
MDL Number: MFCD21605919
CAS Number: 1361114-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR