4-Piperidin-3-yl-6-trifluoromethylpyrimidin-2-ylamine dihydrochloride

Supplier: Matrix Scientific

067635-250MG
101934-388EA 558.46 USD
101934-388
4-Piperidin-3-yl-6-trifluoromethylpyrimidin-2-ylamine dihydrochloride
4-Piperidin-3-yl-6-trifluoromethylpyrimidin-2-ylamine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅Cl₂F₃N₄
MW: 319.16 g/mol
MDL Number: MFCD21605918
CAS Number: 1361111-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR