2-Methyl-4-piperidin-3-yl-6-trifluoromethylpyrimidine hydrochloride
Supplier: Matrix Scientific
067634-250MG
101934-386EA
476.92
USD
101934-386
2-Methyl-4-piperidin-3-yl-6-trifluoromethylpyrimidine hydrochloride
2-Methyl-4-piperidin-3-yl-6-trifluoromethylpyrimidine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₅ClF₃N₃ |
MDL Number:
MFCD21605917 CAS Number: 1361116-99-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...