N-(1-Ethyl-1H-pyrazol-4-yl)-2-piperidin-4-ylacetamide hydrochloride

Supplier: Matrix Scientific

067627-500MG
101934-372EA 623.08 USD
101934-372
N-(1-Ethyl-1H-pyrazol-4-yl)-2-piperidin-4-ylacetamide hydrochloride
N-(1-Ethyl-1H-pyrazol-4-yl)-2-piperidin-4-ylacetamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₁ClN₄O
MW: 272.78 g/mol
MDL Number: MFCD21605911
CAS Number: 1361113-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR