Azetidine-3-carboxylic acid (2-methyl-3H-benzoimidazol-5-yl)amide

Supplier: Matrix Scientific

067625-500MG
101934-368EA 623.08 USD
101934-368
Azetidine-3-carboxylic acid (2-methyl-3H-benzoimidazol-5-yl)amide
Azetidine-3-carboxylic acid (2-methyl-3H-benzoimidazol-5-yl)amide

Hazard: Irritant

Formula: C₁₂H₁₄N₄O
MW: 230.27 g/mol
MDL Number: MFCD21605909
CAS Number: 1361116-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR