1-[4-(4-Piperazin-1-ylphenoxy)piperidin-1-yl]ethanone

Supplier: Matrix Scientific

067611-250MG
101934-340EA 476.92 USD
101934-340
1-[4-(4-Piperazin-1-ylphenoxy)piperidin-1-yl]ethanone
1-[4-(4-Piperazin-1-ylphenoxy)piperidin-1-yl]ethanone

Hazard: Irritant

Formula: C₁₇H₂₅N₃O₂
MDL Number: MFCD16621953
CAS Number: 1361116-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR