1-(3-Trifluoromethylpyridin-2-yl)pyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

067610-250MG
101934-338EA 476.92 USD
101934-338
1-(3-Trifluoromethylpyridin-2-yl)pyrrolidine-3-carboxylic acid
1-(3-Trifluoromethylpyridin-2-yl)pyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₁F₃N₂O₂
MDL Number: MFCD17614588
CAS Number: 1316222-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR