1-Ethanesulfonylpyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

067608-500MG
101934-334EA 623.08 USD
101934-334
1-Ethanesulfonylpyrrolidine-3-carboxylic acid
1-Ethanesulfonylpyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₇H₁₃NO₄S
MDL Number: MFCD17272679
CAS Number: 1316222-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR