[1-(4,6-Dimethylpyrimidin-2-yl)piperidin-4-yl]acetic acid

Supplier: Matrix Scientific

067606-500MG
101934-330EA 623.08 USD
101934-330
[1-(4,6-Dimethylpyrimidin-2-yl)piperidin-4-yl]acetic acid
[1-(4,6-Dimethylpyrimidin-2-yl)piperidin-4-yl]acetic acid

Hazard: Irritant

Formula: C₁₃H₁₉N₃O₂
MDL Number: MFCD16621914
CAS Number: 1361112-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR