2-[5-(3-Aminomethylphenyl)-4-pyridin-4-ylpyrazol-1-yl]ethanol

Supplier: Matrix Scientific

067605-250MG
101934-328EA 476.92 USD
101934-328
2-[5-(3-Aminomethylphenyl)-4-pyridin-4-ylpyrazol-1-yl]ethanol
2-[5-(3-Aminomethylphenyl)-4-pyridin-4-ylpyrazol-1-yl]ethanol

Hazard: Irritant

Formula: C₁₇H₁₈N₄O
MDL Number: MFCD21605896
CAS Number: 1361114-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR