(1-Phenethylpiperidin-4-yl)acetic acid hydrochloride

Supplier: Matrix Scientific

067604-500MG
101934-326EA 623.08 USD
101934-326
(1-Phenethylpiperidin-4-yl)acetic acid hydrochloride
(1-Phenethylpiperidin-4-yl)acetic acid hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClNO₂
MDL Number: MFCD21605895
CAS Number: 1361116-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR