(1-Methanesulfonylpiperidin-4-yl)acetic acid

Supplier: Matrix Scientific

067602-250MG
101934-322EA 400 USD
101934-322
(1-Methanesulfonylpiperidin-4-yl)acetic acid
(1-Methanesulfonylpiperidin-4-yl)acetic acid

Hazard: Irritant

Formula: C₈H₁₅NO₄S
MDL Number: MFCD11585618
CAS Number: 423722-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR