4-(4-Fluorobenzyl)morpholine-2-carboxylic acid

Supplier: Matrix Scientific

067601-250MG
101934-320EA 561.54 USD
101934-320
4-(4-Fluorobenzyl)morpholine-2-carboxylic acid
4-(4-Fluorobenzyl)morpholine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₄FNO₃
MDL Number: MFCD16223993
CAS Number: 1307552-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR