3-Piperidin-4-yl-5,6-dihydro-8H-[1,2,4]triazolo[4,3-a]pyrazine-7-carboxylic acid tert-butyl ester

Supplier: Matrix Scientific

067596-250MG
101934-310EA 638.46 USD
101934-310
3-Piperidin-4-yl-5,6-dihydro-8H-[1,2,4]triazolo[4,3-a]pyrazine-7-carboxylic acid tert-butyl ester
3-Piperidin-4-yl-5,6-dihydro-8H-[1,2,4]triazolo[4,3-a]pyrazine-7-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₅H₂₅N₅O₂
MW: 307.4 g/mol
MDL Number: MFCD21605892
CAS Number: 1361112-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR