3-(2-Dimethylamino-4-morpholin-2-ylpyrimidin-5-yl)benzamide hydrochloride

Supplier: Matrix Scientific

067593-250MG
101934-304EA 476.92 USD
101934-304
3-(2-Dimethylamino-4-morpholin-2-ylpyrimidin-5-yl)benzamide hydrochloride
3-(2-Dimethylamino-4-morpholin-2-ylpyrimidin-5-yl)benzamide hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₂ClN₅O₂
MDL Number: MFCD21605890
CAS Number: 1361115-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR