(3aS,9aS)-3a-Methyl-4-(3-trifluoromethylbenzyl)decahydro-1,4-diazacyclopentacycloocten-5-one

Supplier: Matrix Scientific

067591-250MG
101934-300EA 687.69 USD
101934-300
(3aS,9aS)-3a-Methyl-4-(3-trifluoromethylbenzyl)decahydro-1,4-diazacyclopentacycloocten-5-one
(3aS,9aS)-3a-Methyl-4-(3-trifluoromethylbenzyl)decahydro-1,4-diazacyclopentacycloocten-5-one

Hazard: Irritant

Formula: C₁₈H₂₃F₃N₂O
MDL Number: MFCD19691655
CAS Number: 1442080-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR